2-Cyano-6-methylpyridine, 97%, Thermo Scientific Chemicals
2-Cyano-6-methylpyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Cyano-6-methylpyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 425920050
Price (USD)/ Each
122.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
122.00
Add to cart
2-Cyano-6-methylpyridine, 97%, Thermo Scientific Chemicals
Catalog number425920050
Price (USD)/ Each
122.00
-
Add to cart
Chemical Identifiers
CAS1620-75-3
IUPAC Name6-methylpyridine-2-carbonitrile
Molecular FormulaC7H6N2
InChI KeyCMADFEQMYFNYCF-UHFFFAOYSA-N
SMILESCC1=CC=CC(=N1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=96 %
Appearance (Color)White to orange to brown
Melting point68°C to 73°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only