Suberic acid bis(N-hydroxysuccinimide ester), 97%, Thermo Scientific Chemicals
Suberic acid bis(N-hydroxysuccinimide ester), 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Suberic acid bis(N-hydroxysuccinimide ester), 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 426020050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS68528-80-3
IUPAC Namebis(2,5-dioxopyrrolidin-1-yl) octanedioate
Molecular FormulaC16H20N2O8
InChI KeyZWIBGKZDAWNIFC-UHFFFAOYSA-N
SMILESO=C(CCCCCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
NMR (1H-NMR)Consistent with structure
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only