Thermo Scientific Chemicals

2,3,6-Trifluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 426030010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2,3,6-Trifluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 426030010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 426030010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
114152-23-7
IUPAC Name
2-(2,3,6-trifluorophenyl)acetic acid
Molecular Formula
C8H5F3O2
InChI Key
QRAZASHLGLHKEB-UHFFFAOYSA-N
SMILES
OC(=O)CC1=C(F)C=CC(F)=C1F
Appearance (Form)
Crystalline powder or solid
NMR (1H-NMR)
Consistent with structure
HPLC
>=97.5 %
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text