Thermo Scientific Chemicals

Tebbe reagent, 0.5M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 426061000
100 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

Tebbe reagent, 0.5M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 426061000
100 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 426061000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
67719-69-1
IUPAC Name
titanium(4+) (dimethylalumanyl)methanide bis(cyclopenta-2,4-dien-1-ide) chloride
Molecular Formula
C13H18AlClTi
InChI Key
HIVIAXIRSNAVAD-UHFFFAOYSA-M
SMILES
[Cl-].[Ti+4].C[Al](C)[CH2-].[CH-]1C=CC=C1.[CH-]1C=CC=C1
Appearance (Form)
Solution
Appearance (Color)
Dark red to purple
Strength
0.40 to 0.60 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination), Lewis acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text