Thermo Scientific Chemicals

1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 426078000
800 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 426078000
800 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 426078000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16466-97-0
IUPAC Name
bromo(prop-1-yn-1-yl)magnesium
Molecular Formula
C3H3BrMg
InChI Key
YVJBCTFVENNIOV-UHFFFAOYSA-M
SMILES
CC#C[Mg]Br
Appearance (Color)
Clear colorless to yellow to brown
Appearance (Form)
Solution
Appearance
May develop some turbidity or precipitate
Titration Grignard
0.40 to 0.60 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text