Thermo Scientific Chemicals

Ammonium sulfide, 20% solution in water, Thermo Scientific Chemicals

Catalog number: 426080010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Ammonium sulfide, 20% solution in water, Thermo Scientific Chemicals

Catalog number: 426080010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 426080010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12135-76-1
IUPAC Name
diamine sulfane
Molecular Formula
H8N2S
InChI Key
UXXGWUQNRMPNKH-UHFFFAOYSA-N
SMILES
N.N.S
Titration Iodimetric
20.0 to 24.0 %
Appearance (Form)
Solution
Appearance (Color)
Clear yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text