Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-glutamic acid gamma-tert-butyl ester, 98+%, Thermo Scientific Chemicals

Catalog number: 426150050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-glutamic acid gamma-tert-butyl ester, 98+%, Thermo Scientific Chemicals

Catalog number: 426150050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 426150050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3886-08-6
IUPAC Name
(2S)-2-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoate
Molecular Formula
C17H22NO6
InChI Key
GLMODRZPPBZPPB-ZDUSSCGKSA-M
SMILES
CC(C)(C)OC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O
Infrared spectrum
Conforms
Melting point
86°C to 91°C
HPLC
>=97.5 %
Appearance (Color)
White to off-white
NMR (1H-NMR)
Consistent with structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text