N-BOC-1,4-butanediamine, 95%, Thermo Scientific Chemicals
N-BOC-1,4-butanediamine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-BOC-1,4-butanediamine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
1 mL
Packaging:
Glass Bottle
Catalog number 426190050
Price (USD)/ Each
354.00
-
Add to cart
Quantity:
5 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
354.00
Add to cart
N-BOC-1,4-butanediamine, 95%, Thermo Scientific Chemicals
Catalog number426190050
Price (USD)/ Each
354.00
-
Add to cart
Chemical Identifiers
CAS68076-36-8
IUPAC Nametert-butyl N-(4-azaniumylbutyl)carbamate
Molecular FormulaC9H21N2O2
InChI KeyZFQWJXFJJZUVPI-UHFFFAOYSA-O
SMILESCC(C)(C)OC(=O)NCCCC[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=94 %
NMRconsistent with structure
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only