Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Replace glass and metal lab funnels with Thermo Scientific™ Nalgene™ Large HDPE Funnel, featuring external and internal ribbing for air release. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are both shatterproof and dentproof making them a valuble asset to lab safety programs.
Diameter (Metric)Top | 201mm |
Capacity (Metric) | 1.9 L |
Diameter (Metric) Stem | 27 mm |
Material | HDPE |
Height (Metric) | 246 mm |
Autoclavable | Not Autoclavable |
Description | Funnel Large HDPE 203mm |
Length (Metric) Stem | 88 mm |
Quantity | 1/Pk.; 6/Cs. |
Unit Size | Case of 6 |
Diameter (Metric)Top | 201mm |
Capacity (Metric) | 1.9 L |
Diameter (Metric) Stem | 27 mm |
Material | HDPE |
Height (Metric) | 246 mm |
Autoclavable | Not Autoclavable |
Description | Funnel Large HDPE 203mm |
Length (Metric) Stem | 88 mm |
Quantity | 1/Pk.; 6/Cs. |
Unit Size | Each |
Diameter (Metric)Top | 258mm |
Capacity (Metric) | 4.06 L |
Diameter (Metric) Stem | 25 mm |
Material | HDPE |
Height (Metric) | 301 mm |
Autoclavable | Not Autoclavable |
Description | Funnel Large HDPE 279mm |
Length (Metric) Stem | 90 mm |
Quantity | 1/Pk.; 6/Cs. |
Unit Size | Case of 6 |
Diameter (Metric)Top | 258mm |
Capacity (Metric) | 4.06 L |
Diameter (Metric) Stem | 25 mm |
Material | HDPE |
Height (Metric) | 301 mm |
Autoclavable | Not Autoclavable |
Description | Funnel Large HDPE 279mm |
Length (Metric) Stem | 90 mm |
Quantity | 1/Pk.; 6/Cs. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Diameter (Metric)Top | Capacity (Metric) | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
4262-0080 | Case of 6 | 201mm | 1.9 L | 196.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4262-0080PK also known as 4262-0080 | Each | 201mm | 1.9 L | 38.40 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4262-0110 | Case of 6 | 258mm | 4.06 L | 455.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4262-0110PK also known as 4262-0110 | Each | 258mm | 4.06 L | 89.80 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|