Thermo Scientific Chemicals

Isoindoline, 97%, Thermo Scientific Chemicals

Catalog number: 426280050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Isoindoline, 97%, Thermo Scientific Chemicals

Catalog number: 426280050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 426280050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
496-12-8
IUPAC Name
2,3-dihydro-1H-isoindole
Molecular Formula
C8H9N
InChI Key
GWVMLCQWXVFZCN-UHFFFAOYSA-N
SMILES
C1NCC2=CC=CC=C12
Appearance (Form)
Liquid
Refractive index
1.5670 to 1.5710 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow to pink to orange-brown
GC
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text