Thermo Scientific Chemicals

2-Bromo-6-(hydroxymethyl)pyridine, 96%, Thermo Scientific Chemicals

Catalog number: 426310010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Bromo-6-(hydroxymethyl)pyridine, 96%, Thermo Scientific Chemicals

Catalog number: 426310010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 426310010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33674-96-3
IUPAC Name
(6-bromopyridin-2-yl)methanol
Molecular Formula
C6H6BrNO
InChI Key
XDDGKNRSCDEWBR-UHFFFAOYSA-N
SMILES
OCC1=CC=CC(Br)=N1
Appearance (Color)
White to yellow to light brown
Appearance (Form)
Solid
Infrared spectrum
Conforms
NMR (1H-NMR)
Consistent with structure
Melting point
34°C to 41°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text