2-Bromo-6-(hydroxymethyl)pyridine, 96%
2-Bromo-6-(hydroxymethyl)pyridine, 96%
2-Bromo-6-(hydroxymethyl)pyridine, 96%
2-Bromo-6-(hydroxymethyl)pyridine, 96%
Thermo Scientific Chemicals

2-Bromo-6-(hydroxymethyl)pyridine, 96%

CAS: 33674-96-3 | C6H6BrNO | 188.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 426310050
Price (EUR)
171,70
Online Exclusive
207,00
Save 35,30 (17%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
171,70
Online Exclusive
207,00
Save 35,30 (17%)
Each
Add to cart
Chemical Identifiers
CAS33674-96-3
IUPAC Name(6-bromopyridin-2-yl)methanol
Molecular FormulaC6H6BrNO
InChI KeyXDDGKNRSCDEWBR-UHFFFAOYSA-N
SMILESOCC1=CC=CC(Br)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
NMR (1H-NMR)Consistent with structure
Melting point34°C to 41°C
GC>=95 %
Appearance (Color)White to yellow to light brown
Appearance (Form)Solid
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
339568Certificate of AnalysisNov 08, 2022426310050, 426310010
1 result displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P312 - Call a POISON CENTER or doctor if you feel unwell

P332 + P313 - If skin irritation occurs: Get medical advice/attention

P337 + P313 - If eye irritation persists: Get medical advice/attention

Share catalog number, name or link

1x1 image pixel for data collection