Indan-2-carboxylic acid, 98%, Thermo Scientific Chemicals
Indan-2-carboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indan-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
View all
Glass bottle
Catalog number 426360010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS25177-85-9
IUPAC Name2,3-dihydro-1H-indene-2-carboxylic acid
Molecular FormulaC10H10O2
InChI KeyXUDCMQBOWOLYCF-UHFFFAOYSA-N
SMILESOC(=O)C1CC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Crystalline powder
Melting point128°C to 132°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only