Thermo Scientific Chemicals

Methyl 4-iodobutyrate, 95%, stabilized, Thermo Scientific Chemicals

Catalog number: 426370050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 4-iodobutyrate, 95%, stabilized, Thermo Scientific Chemicals

Catalog number: 426370050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 426370050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14273-85-9
IUPAC Name
methyl 4-iodobutanoate
Molecular Formula
C5H9IO2
InChI Key
NBCIIVXSBPDKOM-UHFFFAOYSA-N
SMILES
COC(=O)CCCI
NMR (1H-NMR)
Consistent with structure
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Stabilizer
Copper
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text