DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
Thermo Scientific Chemicals

DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water

CAS: 3483-12-3 | C4H10O2S2 | 154.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
250 mL
Catalog number 426382500
Price (EUR)
336,60
Online Exclusive
396,00
Save 59,40 (15%)
Each
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
336,60
Online Exclusive
396,00
Save 59,40 (15%)
Each
Add to cart
Chemical Identifiers
CAS3483-12-3
IUPAC Name(2R,3R)-1,4-disulfanylbutane-2,3-diol
Molecular FormulaC4H10O2S2
InChI KeyVHJLVAABSRFDPM-IMJSIDKUSA-N
SMILESO[C@@H](CS)[C@@H](O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
UVat 500 nm A: =<0.05
Titration Iodimetric0.90 to 1.10 M
UV(neat)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only

Share catalog number, name or link