2-(Diphenylphosphino)benzaldehyde, 97%, Thermo Scientific Chemicals
2-(Diphenylphosphino)benzaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Diphenylphosphino)benzaldehyde, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
Catalog number 426400010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-(diphenylphosphanyl)benzaldehyde
Molecular FormulaC19H15OP
InChI KeyDRCPJRZHAJMWOU-UHFFFAOYSA-N
SMILESO=CC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)290.30
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point112°C to 120°C
Appearance (Form)Crystalline powder
GC>=96.0 %
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only