Thermo Scientific Chemicals

L-Tyrosine disodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 426410250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

L-Tyrosine disodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 426410250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 426410250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69847-45-6
IUPAC Name
disodium (2S)-2-amino-3-(4-oxidophenyl)propanoate
Molecular Formula
C9H9NNa2O3
InChI Key
ASIYFCYUCMQNGK-RIYJSUBBNA-L
SMILES
[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O
Appearance (Color)
White to tan
Infrared spectrum
Conforms
Specific optical rotation
-9° to -14.5° (20°C, 589 nm) (c=4 in H2O)
Appearance (Form)
Crystalline powder
Water
=<19.3 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text