Thermo Scientific Chemicals

Iodotrimethylsilane, 95-97%, stabilized, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 426421000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Iodotrimethylsilane, 95-97%, stabilized, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 426421000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 426421000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16029-98-4
IUPAC Name
iodotrimethylsilane
Molecular Formula
C3H9ISi
InChI Key
CSRZQMIRAZTJOY-UHFFFAOYSA-N
SMILES
C[Si](C)(C)I
Stabilizer
Copper
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to red
Infrared spectrum
Conforms
Titration Argentometric
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text