Thermo Scientific Chemicals

Chlorotrimethylsilane, 98%, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 426431000
100 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

Chlorotrimethylsilane, 98%, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 426431000
100 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 426431000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-77-4
IUPAC Name
chlorotrimethylsilane
Molecular Formula
C3H9ClSi
InChI Key
IJOOHPMOJXWVHK-UHFFFAOYSA-N
SMILES
C[Si](C)(C)Cl
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Color)
Clear colorless to slightly yellow
Refractive index
1.3865 to 1.3905 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text