Thermo Scientific Chemicals

2-Cyclopentylphenol, 98+%, Thermo Scientific Chemicals

Catalog number: 426510010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Cyclopentylphenol, 98+%, Thermo Scientific Chemicals

Catalog number: 426510010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 426510010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1518-84-9
IUPAC Name
2-cyclopentylphenol
Molecular Formula
C11H14O
InChI Key
JHEKSKQMOBLXQS-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1C1CCCC1
Appearance (Color)
White to orange
Appearance (Form)
Low melting solid
NMR (1H-NMR)
Consistent with structure
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text