2-Cyclopentylphenol, 98+%, Thermo Scientific Chemicals
2-Cyclopentylphenol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Cyclopentylphenol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 426510050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1518-84-9
IUPAC Name2-cyclopentylphenol
Molecular FormulaC11H14O
InChI KeyJHEKSKQMOBLXQS-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1C1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to orange
Appearance (Form)Low melting solid
NMR (1H-NMR)Consistent with structure
GC>=97.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only