Thermo Scientific Chemicals

Methyl undecanoate, 97%, Thermo Scientific Chemicals

Catalog number: 426580250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl undecanoate, 97%, Thermo Scientific Chemicals

Catalog number: 426580250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 426580250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1731-86-8
IUPAC Name
methyl undecanoate
Molecular Formula
C12H24O2
InChI Key
XPQPWPZFBULGKT-UHFFFAOYSA-N
SMILES
CCCCCCCCCCC(=O)OC
Appearance (Form)
Liquid
Infrared spectrum
Conforms
NMR (1H-NMR)
Consistent with structure
GC
>=96.0 %
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text