Thermo Scientific Chemicals

3-(Trimethoxysilyl)propyl acrylate, 90%, stabilized, Thermo Scientific Chemicals

Catalog number: 426600250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-(Trimethoxysilyl)propyl acrylate, 90%, stabilized, Thermo Scientific Chemicals

Catalog number: 426600250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 426600250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4369-14-6
IUPAC Name
3-(trimethoxysilyl)propyl prop-2-enoate
Molecular Formula
C9H18O5Si
InChI Key
KBQVDAIIQCXKPI-UHFFFAOYSA-N
SMILES
CO[Si](CCCOC(=O)C=C)(OC)OC
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.4280 to 1.4300 (20°C, 589 nm)
GC
>=90 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text