Thermo Scientific Chemicals

Bromoacetic anhydride, 95%, Thermo Scientific Chemicals

Catalog number: 426700050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Bromoacetic anhydride, 95%, Thermo Scientific Chemicals

Catalog number: 426700050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 426700050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13094-51-4
IUPAC Name
2-bromoacetyl 2-bromoacetate
Molecular Formula
C4H4Br2O3
InChI Key
FUKOTTQGWQVMQB-UHFFFAOYSA-N
SMILES
BrCC(=O)OC(=O)CBr
Infrared spectrum
Conforms
Melting point
34°C to 42°C
Titration Argentometric
94 to 106 % (ex Br)
Appearance (Color)
White to brown
Appearance (Form)
Solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text