3,3-Diethoxy-1-propanol, 98%, Thermo Scientific Chemicals
3,3-Diethoxy-1-propanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3-Diethoxy-1-propanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 426720010
Price (USD)/ Each
54.60
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
54.60
Add to cart
3,3-Diethoxy-1-propanol, 98%, Thermo Scientific Chemicals
Catalog number426720010
Price (USD)/ Each
54.60
-
Add to cart
Chemical Identifiers
IUPAC Name3,3-diethoxypropan-1-ol
Molecular FormulaC7H16O3
InChI KeyASERXEZXVIJBRO-UHFFFAOYSA-N
SMILESCCOC(CCO)OCC
Molecular Weight (g/mol)148.20
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only