Thermo Scientific Chemicals

3-Methoxyphenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 426758000
800 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

3-Methoxyphenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 426758000
800 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 426758000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
36282-40-3
IUPAC Name
bromo(3-methoxyphenyl)magnesium
Molecular Formula
C7H7BrMgO
InChI Key
AJKNYEHIJAMGHX-UHFFFAOYSA-M
SMILES
COC1=CC=CC([Mg]Br)=C1
Appearance
May develop some turbidity or precipitate
Appearance (Color)
Clear yellow-brown
Titration Grignard
0.9 to 1.1 M
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text