Thermo Scientific Chemicals

p-Tolylmagnesium bromide, approx. 0.5M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 426768000
800 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

p-Tolylmagnesium bromide, approx. 0.5M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 426768000
800 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 426768000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4294-57-9
IUPAC Name
bromo(4-methylphenyl)magnesium
Molecular Formula
C7H7BrMg
InChI Key
ZRJNGFJIBZKXTP-UHFFFAOYSA-M
SMILES
CC1=CC=C([Mg]Br)C=C1
Appearance
May develop some turbidity or precipitate
Appearance (Form)
Solution
Appearance (Color)
Clear to hazy yellow to amber
Titration Grignard
0.40 to 0.60 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text