Thermo Scientific Chemicals

(4-Bromophenoxy)-tert-butyldimethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: 427090250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

(4-Bromophenoxy)-tert-butyldimethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: 427090250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 427090250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
67963-68-2
IUPAC Name
(4-bromophenoxy)(tert-butyl)dimethylsilane
Molecular Formula
C12H19BrOSi
InChI Key
DLGZGLKSNRKLSM-UHFFFAOYSA-N
SMILES
CC(C)(C)[Si](C)(C)OC1=CC=C(Br)C=C1
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow
NMR (1H-NMR)
Consistent with structure
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text