Triisopropyl borate, 98+%, AcroSeal™, Thermo Scientific Chemicals
Triisopropyl borate, 98+%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triisopropyl borate, 98+%, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 427148000
Price (USD)/ Each
-
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS5419-55-6
IUPAC Nametris(propan-2-yl) borate
Molecular FormulaC9H21BO3
InChI KeyNHDIQVFFNDKAQU-UHFFFAOYSA-N
SMILESCC(C)OB(OC(C)C)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=98.0 %
Refractive index1.3755 to 1.3775 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only