Ethanolamine, 99%, AcroSeal™, Thermo Scientific Chemicals
Ethanolamine, 99%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethanolamine, 99%, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number 427251000
Price (USD)/ Each
72.65
Online exclusive
80.40 
Save 7.75 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)/ Each
72.65
Online exclusive
80.40 
Save 7.75 (10%)
Add to cart
Ethanolamine, 99%, AcroSeal™, Thermo Scientific Chemicals
Catalog number427251000
Price (USD)/ Each
72.65
Online exclusive
80.40 
Save 7.75 (10%)
-
Add to cart
Chemical Identifiers
CAS141-43-5
IUPAC Name2-aminoethan-1-ol
Molecular FormulaC2H7NO
InChI KeyHZAXFHJVJLSVMW-UHFFFAOYSA-N
SMILESNCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4530 to 1.4550 (20°C, 589 nm)
Color scale=<15 APHA
Infrared spectrumConforms
GC>=98.5 %
Water=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only