Chlorodiisopropylphosphine, 96%, AcroSeal™, Thermo Scientific Chemicals
Chlorodiisopropylphosphine, 96%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chlorodiisopropylphosphine, 96%, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number 427270500
Price (USD)/ Each
315.65
Online exclusive
351.00 
Save 35.35 (10%)
-
Add to cart
Quantity:
50 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)/ Each
315.65
Online exclusive
351.00 
Save 35.35 (10%)
Add to cart
Chlorodiisopropylphosphine, 96%, AcroSeal™, Thermo Scientific Chemicals
Catalog number427270500
Price (USD)/ Each
315.65
Online exclusive
351.00 
Save 35.35 (10%)
-
Add to cart
Chemical Identifiers
CAS40244-90-4
IUPAC Namechlorobis(propan-2-yl)phosphane
Molecular FormulaC6H14ClP
InChI KeyJZPDBTOWHLZQFC-UHFFFAOYSA-N
SMILESCC(C)P(Cl)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4740 to 1.4760 (20°C, 589 nm)
Appearance (Color)Clear light yellow
Infrared spectrumConforms
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only