3,5-Dimethoxybenzyl bromide, 95%, Thermo Scientific Chemicals
3,5-Dimethoxybenzyl bromide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dimethoxybenzyl bromide, 95%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 427340050
Price (USD)/ Each
94.90
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
94.90
Add to cart
3,5-Dimethoxybenzyl bromide, 95%, Thermo Scientific Chemicals
Catalog number427340050
Price (USD)/ Each
94.90
-
Add to cart
Chemical Identifiers
CAS877-88-3
IUPAC Name1-(bromomethyl)-3,5-dimethoxybenzene
Molecular FormulaC9H11BrO2
InChI KeyBTHIGJGJAPYFSJ-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=CC(CBr)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point68°C to 72°C
Appearance (Color)White to brown
Infrared spectrumConforms
Appearance (Form)Powder or crystals and/or chunks
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only