Isobutylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Isobutylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isobutylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number 427391000
Price (USD)/ Each
88.65
Online exclusive
97.90 
Save 9.25 (9%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)/ Each
88.65
Online exclusive
97.90 
Save 9.25 (9%)
Add to cart
Isobutylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Catalog number427391000
Price (USD)/ Each
88.65
Online exclusive
97.90 
Save 9.25 (9%)
-
Add to cart
Chemical Identifiers
IUPAC Namebromo(2-methylpropyl)magnesium
Molecular FormulaC4H9BrMg
InChI KeyCMWBEISSZHZIMU-UHFFFAOYSA-M
SMILESCC(C)C[Mg]Br
Molecular Weight (g/mol)161.33
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Grignard1.9 to 2.2 M
Appearance (Form)Solution
Appearance (Color)Yellow to dark brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only