Thermo Scientific Chemicals

Isobutylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 427398000
800 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

Isobutylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 427398000
800 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 427398000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
926-62-5
IUPAC Name
bromo(2-methylpropyl)magnesium
Molecular Formula
C4H9BrMg
InChI Key
CMWBEISSZHZIMU-UHFFFAOYSA-M
SMILES
CC(C)C[Mg]Br
Appearance (Form)
Solution
Titration Grignard
1.9 to 2.2 M
Appearance (Color)
Yellow to dark brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text