Methylmagnesium iodide, 3M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Methylmagnesium iodide, 3M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methylmagnesium iodide, 3M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 427401000
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.70 
Save 11.05 (14%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.70 
Save 11.05 (14%)
Add to cart
Methylmagnesium iodide, 3M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Catalog number427401000
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.70 
Save 11.05 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Nameiodo(methyl)magnesium
Molecular FormulaCH3IMg
InChI KeyAUPXBVDHVRZMIB-UHFFFAOYSA-M
SMILESC[Mg]I
Molecular Weight (g/mol)166.24
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear to hazy solution
Strength2.85 to 3.50 M
Appearance (Color)Yellow to grey to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only