3,5-Difluorophenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
3,5-Difluorophenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Difluorophenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 427441000
Price (USD)/ Each
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namebromo(3,5-difluorophenyl)magnesium
Molecular FormulaC6H3BrF2Mg
InChI KeyCURGXPMIRCWEDB-UHFFFAOYSA-M
SMILESFC1=CC([Mg]Br)=CC(F)=C1
Molecular Weight (g/mol)217.30
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green or brown
Appearance (Form)Solution
Titration Grignard0.40 to 0.75 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only