Thermo Scientific Chemicals

Cyclopentylmagnesium bromide, 2.0M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 427458000
800 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Cyclopentylmagnesium bromide, 2.0M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 427458000
800 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 427458000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33240-34-5
IUPAC Name
bromo(cyclopentyl)magnesium
Molecular Formula
C5H9BrMg
InChI Key
PZOJUJCALDFFHC-UHFFFAOYSA-M
SMILES
Br[Mg]C1CCCC1
Appearance (Color)
Grey
Appearance (Form)
Solution
Titration Grignard
1.90 to 2.20 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text