Thermo Scientific Chemicals

Isovaleronitrile, 98%, Thermo Scientific Chemicals

Catalog number: 427560050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Isovaleronitrile, 98%, Thermo Scientific Chemicals

Catalog number: 427560050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 427560050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
625-28-5
IUPAC Name
3-methylbutanenitrile
Molecular Formula
C5H9N
InChI Key
QHDRKFYEGYYIIK-UHFFFAOYSA-N
SMILES
CC(C)CC#N
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.3920 to 1.3940 (20°C, 589 nm)
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text