Isovaleronitrile, 98%, Thermo Scientific Chemicals
Isovaleronitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isovaleronitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 427560250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS625-28-5
IUPAC Name3-methylbutanenitrile
Molecular FormulaC5H9N
InChI KeyQHDRKFYEGYYIIK-UHFFFAOYSA-N
SMILESCC(C)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3920 to 1.3940 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only