Thermo Scientific Chemicals

DL-Pantolactone, 97%, Thermo Scientific Chemicals

Catalog number: 427591000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-Pantolactone, 97%, Thermo Scientific Chemicals

Catalog number: 427591000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 427591000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-50-5
IUPAC Name
3-hydroxy-4,4-dimethyloxolan-2-one
Molecular Formula
C6H10O3
InChI Key
SERHXTVXHNVDKA-UHFFFAOYNA-N
SMILES
CC1(C)COC(=O)C1O
Water
=<0.5 %
Appearance (Color)
White to light yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text