Isophorone diisocyanate, 98+%, mixture of isomers
Isophorone diisocyanate, 98+%, mixture of isomers
Isophorone diisocyanate, 98+%, mixture of isomers
Isophorone diisocyanate, 98+%, mixture of isomers
Isophorone diisocyanate, 98+%, mixture of isomers
Thermo Scientific Chemicals

Isophorone diisocyanate, 98+%, mixture of isomers

CAS: 4098-71-9 | C12H18N2O2 | 222.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 427600010
Price (EUR)
140,25
Online Exclusive
165,00
Save 24,75 (15%)
Each
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
140,25
Online Exclusive
165,00
Save 24,75 (15%)
Each
Add to cart
Chemical Identifiers
CAS4098-71-9
IUPAC Name5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
Molecular FormulaC12H18N2O2
InChI KeyNIMLQBUJDJZYEJ-UHFFFAOYNA-N
SMILESCC1(C)CC(CC(C)(CN=C=O)C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4830 to 1.4850 (20°C, 589 nm)
GC>=98.0 % (sum of mixed isomers)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473282Certificate of AnalysisJun 06, 2025427600500, 427600010, 427602500
A0469675Certificate of AnalysisFeb 06, 2025427600500, 427600010, 427602500
A0460580Certificate of AnalysisFeb 26, 2024427600500, 427600010, 427602500
A0456517Certificate of AnalysisSep 21, 2023427600500, 427600010, 427602500
A0437506Certificate of AnalysisAug 25, 2023427600500, 427600010, 427602500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute Inhalation Toxicity - Vapors
Category 1
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Respiratory Sensitization
Category 1
Skin Sensitization
Category 1
Specific target organ toxicity - (single exposure)
Category 3
Chronic aquatic toxicity
Category 2
Label Elements
Signal Word

Danger

Hazard Statements

H315 - Causes skin irritation

H317 - May cause an allergic skin reaction

H319 - Causes serious eye irritation

H330 - Fatal if inhaled

H334 - May cause allergy or asthma symptoms or breathing difficulties if inhaled

H335 - May cause respiratory irritation

H411 - Toxic to aquatic life with long lasting effects

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P332 + P313 - If skin irritation occurs: Get medical advice/attention

P333 + P313 - If skin irritation or rash occurs: Get medical advice/attention

P337 + P313 - If eye irritation persists: Get medical advice/attention

Share catalog number, name or link

1x1 image pixel for data collection