Thermo Scientific Chemicals

Isophorone diisocyanate, 98%, Thermo Scientific Chemicals

Catalog number: 427600500
50 mL, Glass bottle, Each
Thermo Scientific Chemicals

Isophorone diisocyanate, 98%, Thermo Scientific Chemicals

Catalog number: 427600500
50 mL, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 427600500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4098-71-9
IUPAC Name
5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
Molecular Formula
C12H18N2O2
InChI Key
NIMLQBUJDJZYEJ-UHFFFAOYNA-N
SMILES
CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4830 to 1.4850 (20°C, 589 nm)
GC
>=98.0 % (sum of mixed isomers)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text