Isophorone diisocyanate, 98%, Thermo Scientific Chemicals
Isophorone diisocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isophorone diisocyanate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
1 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 427602500
Price (USD)/ Each
64.65
Online exclusive
72.10 
Save 7.45 (10%)
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
64.65
Online exclusive
72.10 
Save 7.45 (10%)
Add to cart
Isophorone diisocyanate, 98%, Thermo Scientific Chemicals
Catalog number427602500
Price (USD)/ Each
64.65
Online exclusive
72.10 
Save 7.45 (10%)
-
Add to cart
Chemical Identifiers
CAS4098-71-9
IUPAC Name5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
Molecular FormulaC12H18N2O2
InChI KeyNIMLQBUJDJZYEJ-UHFFFAOYNA-N
SMILESCC1(C)CC(CC(C)(CN=C=O)C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4830 to 1.4850 (20°C, 589 nm)
GC>=98.0 % (sum of mixed isomers)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only