Thermo Scientific Chemicals

cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%

Catalog number: 427640010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%

Catalog number: 427640010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 427640010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6217-54-5
IUPAC Name
(4Z,7Z,10Z,13Z,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid
Molecular Formula
C22H32O2
InChI Key
MBMBGCFOFBJSGT-KUBAVDMBSA-N
SMILES
CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCC(O)=O
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Viscous liquid
Infrared spectrum
Conforms
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text