cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%
cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%
Thermo Scientific Chemicals

cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Packaging:
View all
Glass Bottle
Catalog number 427641000
Price (USD)/ Each
-
Quantity:
100 mg
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(4Z,7Z,10Z,13Z,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid
Molecular FormulaC22H32O2
InChI KeyMBMBGCFOFBJSGT-KUBAVDMBSA-N
SMILESCC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCC(O)=O
Molecular Weight (g/mol)328.50
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only