Thermo Scientific Chemicals

Diphenyl-2-pyridylphosphine, 97%, Thermo Scientific Chemicals

Catalog number: 427660010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Diphenyl-2-pyridylphosphine, 97%, Thermo Scientific Chemicals

Catalog number: 427660010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 427660010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
37943-90-1
IUPAC Name
2-(diphenylphosphanyl)pyridine
Molecular Formula
C17H14NP
InChI Key
SVABQOITNJTVNJ-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=N1
Appearance (Color)
White to yellow to orange
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Elemental Analysis
5.17 to 5.55 % H
Elemental Analysis
5.1 to 5.6 % N

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text