Thermo Scientific Chemicals

5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate, 96%, Thermo Scientific Chemicals

Catalog number: 427670010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate, 96%, Thermo Scientific Chemicals

Catalog number: 427670010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 427670010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
129946-88-9
IUPAC Name
8-(trifluoromethyl)-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaen-8-ium trifluoromethanesulfonate
Molecular Formula
C14H8F6O3S2
InChI Key
QXXHXTRTGZBOGD-UHFFFAOYSA-M
SMILES
[O-]S(=O)(=O)C(F)(F)F.FC(F)(F)[S+]1C2=CC=CC=C2C2=CC=CC=C12
Appearance (Color)
White to light brown
Appearance (Form)
Crystalline powder or solid
Infrared spectrum
Conforms
NMR (1H-NMR)
Consistent with structure
HPLC
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text