Thermo Scientific Chemicals

DL-3-Phenyllactic acid, 97%, Thermo Scientific Chemicals

Catalog number: 427770100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-3-Phenyllactic acid, 97%, Thermo Scientific Chemicals

Catalog number: 427770100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 427770100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
828-01-3
IUPAC Name
2-hydroxy-3-phenylpropanoic acid
Molecular Formula
C9H10O3
InChI Key
VOXXWSYKYCBWHO-UHFFFAOYNA-N
SMILES
OC(CC1=CC=CC=C1)C(O)=O
HPLC
>=96.0 %
Appearance (Color)
White to almost white
Appearance (Form)
Powder or crystalline powder
Infrared spectrum
Conforms
NMR (1H-NMR)
Consistent with structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text