Thermo Scientific Chemicals

Bromoacetic acid N-hydroxysuccinimide ester, 90%, Thermo Scientific Chemicals

Catalog number: 427780050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Bromoacetic acid N-hydroxysuccinimide ester, 90%, Thermo Scientific Chemicals

Catalog number: 427780050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 427780050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
42014-51-7
IUPAC Name
2,5-dioxopyrrolidin-1-yl 2-bromoacetate
Molecular Formula
C6H6BrNO4
InChI Key
NKUZQMZWTZAPSN-UHFFFAOYSA-N
SMILES
BrCC(=O)ON1C(=O)CCC1=O
Infrared spectrum
Conforms
Appearance (Color)
White to off-white
Elemental Analysis
>=88 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text