Thermo Scientific Chemicals

Octylmagnesium bromide, 2M solution in diethyl ether,AcroSeal™, Thermo Scientific Chemicals

Catalog number: 427811000
100 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

Octylmagnesium bromide, 2M solution in diethyl ether,AcroSeal™, Thermo Scientific Chemicals

Catalog number: 427811000
100 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 427811000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17049-49-9
IUPAC Name
bromo(octyl)magnesium
Molecular Formula
C8H17BrMg
InChI Key
ZDUZWGUHIRQPGP-UHFFFAOYSA-M
SMILES
CCCCCCCC[Mg]Br
Titration Grignard
1.9 to 2.2 M
Appearance (Color)
Yellow to brown
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text